missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-HT7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP1-46598-0.025ML
This item is not returnable.
View return policy
Description
5-HT7 Polyclonal specifically detects 5-HT7 in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| 5-HT7 | |
| Polyclonal | |
| Unconjugated | |
| Whole antisera with No Preservative | |
| 5-HT-7, 5-HT75-hydroxytryptamine receptor 7, 5-HT-X, 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled), serotonin 5-HT-7 receptor, Serotonin receptor 7 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Specific for 5HT7R receptor. | |
| Mouse, Guinea Pig, Rat, Human | |
| Antisera |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Western Blot | |
| LYOPH | |
| Western Blot 1: 200 - 1: 2000, Immunohistochemistry 1: 200- 1: 2000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1: 200 - 1: 2000 | |
| P34969 | |
| HTR7 | |
| A synthetic peptide from internal region of human 5HT7R receptor (Serotonin receptor 7R) conjugated to an immunogenic carrier protein was used as the antigen. | |
| 0.025 mL | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 3363 | |
| Reconstitute with 0.1 ml sterilized water to desired concentration. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction