missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-HT7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP1-46598
This item is not returnable.
View return policy
Description
5-HT7 Polyclonal specifically detects 5-HT7 in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| 5-HT7 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from whole antisera with No Preservative | |
| 5-HT-7, 5-HT75-hydroxytryptamine receptor 7, 5-HT-X, 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled), serotonin 5-HT-7 receptor, Serotonin receptor 7 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Specific for 5HT7R receptor. | |
| Human, Mouse, Rat, Guinea Pig | |
| Antisera |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:1000 | |
| P34969 | |
| HTR7 | |
| A synthetic peptide from human 5-HT7 conjugated to blue carrier protein was used as the antigen. The peptide is homologous in rat, mouse and guinea pig. | |
| 0.1 mL | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 3363 | |
| Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction