missing translation for 'onlineSavingsMsg'
Learn More

5'-Hydroxy-2'-methoxyacetophenone, TRC

Product Code. 30396380
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396380 250 mg 250mg
30403173 2.5 g 2.5g
30395083 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396380 Supplier Toronto Research Chemicals Supplier No. TRCH806385250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31405-60-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.17
IUPAC Name 1-(5-hydroxy-2-methoxyphenyl)ethanone
SMILES COc1ccc(O)cc1C(=O)C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H10O3/c1-6(10)8-5-7(11)3-4-9(8)12-2/h3-5,11H,1-2H3
Formula Weight 166.063
Chemical Name or Material 5'-Hydroxy-2'-methoxyacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.