missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-N-methylprotriptyline, TRC

Product Code. 30138514
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138514 100 mg 100mg
30011909 10 mg 10mg
30005725 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138514 Supplier Toronto Research Chemicals Supplier No. TRCH948035100MG
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18029-54-4
Molecular Formula C20 H23 N O
Molecular Weight (g/mol) 293.4
Synonym 5H-Dibenzo[a,d]cyclohepten-5-ol, 5-[3-(dimethylamino)propyl]-
IUPAC Name 11-[3-(dimethylamino)propyl]dibenzo[2,1-b:1',2'-e][7]annulen-11-ol
SMILES CN(C)CCCC1(O)c2ccccc2C=Cc3ccccc13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H23NO/c1-21(2)15-7-14-20(22)18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)20/h3-6,8-13,22H,7,14-15H2,1-2H3
Formula Weight 293.178
Chemical Name or Material 5-[3-(Dimethylamino)propyl]-5H-dibenzo[a,d]cyclohepten-5-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.