missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-p-naphthoquinone, 97%

Product Code. 10477500 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10477500 5 g Glass bottle 5g
10132300 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10477500 Supplier Thermo Scientific Acros Supplier No. 121640050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 481-39-0 | C10H6O3 | 174.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 481-39-0
Molecular Formula C10H6O3
Molecular Weight (g/mol) 174.16
MDL Number MFCD00001684
InChI Key KQPYUDDGWXQXHS-UHFFFAOYSA-N
Synonym juglone, 5-hydroxy-1,4-naphthoquinone, 5-hydroxy-1,4-naphthalenedione, regianin, walnut extract, juglon, nucin, 5-hydroxynaphthoquinone, akhnot, yuglon
PubChem CID 3806
ChEBI CHEBI:15794
IUPAC Name 5-hydroxynaphthalene-1,4-dione
SMILES OC1=CC=CC2=C1C(=O)C=CC2=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 161.0°C to 163.0°C
Color Brown to Orange
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 08, 308
Merck Index 15, 5317
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in aq.sol. of alkalies,soluble in alcohol and ether
Formula Weight 174.16
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxy-p-naphthoquinone, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.