missing translation for 'onlineSavingsMsg'
Learn More

5’-Hydroxyequol, TRC

Product Code. 30135283
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135283 25 mg 25mg
30008705 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135283 Supplier Toronto Research Chemicals Supplier No. TRCH82575025MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65998-44-9
Molecular Formula C15H14O4
Molecular Weight (g/mol) 258.27
Synonym Isoflavan-4',5,7-triol, 3,4-dihydro-3-(4-Hydroxyphenyl)-2H-1-benzopyran-5,7-diol, 3-(4-Hydroxyphenyl)chroman-5,7-diol
SMILES OC(C=C1)=CC=C1C2CC3=C(O)C=C(O)C=C3OC2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H14O4/c16-11-3-1-9(2-4-11)10-5-13-14(18)6-12(17)7-15(13)19-8-10/h1-4,6-7,10,16-18H,5,8H2
Formula Weight 258.0892
Chemical Name or Material 5'-Hydroxyequol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.