missing translation for 'onlineSavingsMsg'
Learn More

5-(Hydroxymethyl)-2-iodophenol,97%, Thermo Scientific™

Product Code. 10662275
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10662275 5 g Amber Glass Bottle 5g
10515744 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10662275 Supplier Thermo Scientific Maybridge Supplier No. MO07809DE
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 773869-57-1
Molecular Formula C7H7IO2
Molecular Weight (g/mol) 250.035
InChI Key VIWWSAGABDIIFF-UHFFFAOYSA-N
Synonym 5-hydroxymethyl-2-iodophenol, 3-hydroxy-4-iodobenzyl alcohol, 3-hydroxy-4-iodobenzenemethanol, benzenemethanol,3-hydroxy-4-iodo, zlchem 794, 5-hydroxymethyl-2-iodo-phenol, 3-hydroxy-4-iodophenyl methanol, 5-hydroxymethyl-2-iodanyl-phenol
PubChem CID 18525941
IUPAC Name 5-(hydroxymethyl)-2-iodophenol
SMILES C1=CC(=C(C=C1CO)O)I

Specifications

Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 250.03
Percent Purity 97%
Chemical Name or Material 5-(hydroxymethyl)-2-iodophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.