missing translation for 'onlineSavingsMsg'
Learn More

5-Iodovanillin, 98%

Product Code. 11432158
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432158 250 g 250g
11412158 10 g 10g
11422158 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11432158 Supplier Thermo Scientific Alfa Aesar Supplier No. A15162.30
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5438-36-8 | C8H7IO3 | 278.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5438-36-8
Molecular Formula C8H7IO3
Molecular Weight (g/mol) 278.045
MDL Number MFCD00006941
InChI Key FBBCSYADXYILEH-UHFFFAOYSA-N
Synonym 5-iodovanillin, 5 iodovanillin, benzaldehyde, 4-hydroxy-3-iodo-5-methoxy, 5-iodo vanillin, 3-methoxy-4-hydroxy-5-iodobenzaldehyde, 5-iodovanilline, 5-iodovanilin, 5-iodo vaniline, acmc-1ao68
PubChem CID 79499
IUPAC Name 4-hydroxy-3-iodo-5-methoxybenzaldehyde
SMILES COC1=C(C(=CC(=C1)C=O)I)O

Specifications

Melting Point 181°C to 184°C
Odor Irritating
Quantity 250 g
Beilstein 2049762
Sensitivity Air and light sensitive
Formula Weight 278.05
Percent Purity 98%
Chemical Name or Material 5-Iodovanillin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.