missing translation for 'onlineSavingsMsg'
Learn More

5-Iodovanillin, 98%

Product Code. p-7022402
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11412158

Brand: Thermo Scientific Alfa Aesar A15162.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5438-36-8 | C8H7IO3 | 278.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5438-36-8
Molecular Formula C8H7IO3
Molecular Weight (g/mol) 278.045
MDL Number MFCD00006941
InChI Key FBBCSYADXYILEH-UHFFFAOYSA-N
Synonym 5-iodovanillin, 5 iodovanillin, benzaldehyde, 4-hydroxy-3-iodo-5-methoxy, 5-iodo vanillin, 3-methoxy-4-hydroxy-5-iodobenzaldehyde, 5-iodovanilline, 5-iodovanilin, 5-iodo vaniline, acmc-1ao68
PubChem CID 79499
IUPAC Name 4-hydroxy-3-iodo-5-methoxybenzaldehyde
SMILES COC1=C(C(=CC(=C1)C=O)I)O

Specifications

Melting Point 181°C to 184°C
Odor Irritating
Quantity 10 g
Beilstein 2049762
Sensitivity Air and light sensitive
Formula Weight 278.05
Percent Purity 98%
Chemical Name or Material 5-Iodovanillin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.