missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-methylaniline, 97%, Thermo Scientific™

Product Code. 10516694 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10516694 5 g Glass bottle 5g
10693225 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10516694 Supplier Thermo Scientific Acros Supplier No. 398240050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 50868-72-9
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00075057
InChI Key RPJXLEZOFUNGNZ-UHFFFAOYSA-N
Synonym 5-methoxy-o-toluidine, 2-amino-4-methoxytoluene, benzenamine, 5-methoxy-2-methyl, 5-methoxy-2-methylbenzenamine, 2-methyl-5-ethoxyaniline, 5-methoxy-2-methylphenylamine, 6-methyl-m-anisidine, 3-amino-4-methylanisole 5-methoxy-2-methylaniline, pubchem19827, acmc-1awic
PubChem CID 99500
IUPAC Name 5-methoxy-2-methylaniline
SMILES COC1=CC=C(C)C(N)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 43°C to 46°C
Color White to Yellow
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3OC6H3(CH3)NH2
Quantity 5 g
Formula Weight 137.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 5-Methoxy-2-methylaniline
Show More Show Less
compliance-icons
Product Identifier
  • 5-Methoxy-2-methylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.