missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-nitrophenol, 98%

Product Code. 10249793 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10249793 5 g Glass bottle 5g
10682002 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10249793 Supplier Thermo Scientific Acros Supplier No. 335440050
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 704-14-3 | C7H7NO4 | 169.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 704-14-3
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.14
MDL Number MFCD00100932
InChI Key NRTULWPODYLFOJ-UHFFFAOYSA-N
PubChem CID 219635
IUPAC Name 5-methoxy-2-nitrophenol
SMILES COC1=CC=C(C(O)=C1)[N+]([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 90°C to 94°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Synonym 2-hydroxy-4-methoxynitrobenzene, phenol, 5-methoxy-2-nitro, 2-nitro-5-methoxyphenol, 3-hydroxy-4-nitroanisole, 5-methoxy-2-nitro-phenol, phenol,5-methoxy-2-nitro, pubchem22230, 3-methoxy-6-nitrophenol, 2-hydroxy4-methoxynitrobenzene, 5-methoxy-2-nitrophenol
Solubility Information Solubility in water: .. Other solubilities: soluble in acetone,chloroform,ethyl alcohol
Formula Weight 169.14
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 5-Methoxy-2-nitrophenol
Show More Show Less
compliance-icons
Product Identifier
  • 5-Methoxy-2-nitrophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.