missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3-phenyl-4-isoxazolesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10421303
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10421303 1 g Amber Glass Bottle 1g
10596513 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10421303 Supplier Thermo Scientific Maybridge Supplier No. CC00503DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 857283-56-8
Molecular Formula C10H8ClNO3S
Molecular Weight (g/mol) 257.688
MDL Number MFCD06658966
InChI Key GXQBTNFENYVPFM-UHFFFAOYSA-N
Synonym 5-methyl-3-phenyl-4-isoxazolesulfonyl chloride, 5-methyl-3-phenylisoxazole-4-sulphonyl chloride, 5-methyl-3-phenylisoxazole-4-sulfonyl chloride, chloro 5-methyl-3-phenylisoxazol-4-yl sulfone, 5-methyl-3-phenyl-isoxazole-4-sulfonyl chloride
PubChem CID 18525696
IUPAC Name 5-methyl-3-phenyl-1,2-oxazole-4-sulfonyl chloride
SMILES CC1=C(C(=NO1)C2=CC=CC=C2)S(=O)(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 257.7
Percent Purity 97%
Chemical Name or Material 5-Methyl-3-phenyl-4-isoxazolesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.