missing translation for 'onlineSavingsMsg'
Learn More

5-(Methylthio)thiophene-2-carboxaldehyde, 97%

Product Code. 15468378
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15468378 1 g 1g
15478378 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15468378 Supplier Thermo Scientific Alfa Aesar Supplier No. H61803.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 24445-35-0 | C6H6OS2 | 158.233 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24445-35-0
Molecular Formula C6H6OS2
Molecular Weight (g/mol) 158.233
MDL Number MFCD00173744
InChI Key RKXYTFTZODXDEF-UHFFFAOYSA-N
Synonym 5-methylthio thiophene-2-carbaldehyde, 5-methylthio thiophene-2-carboxaldehyde, 5-methylsulfanyl thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,5-methylthio, 2-formyl-5-methylthiothiphene, 2-formyl-5-methylthio thiophene, 5-methylmercapto-2-thiophenaldehyde, 5-methylthio-2-thiophenecarbaldehyde, 5-methylthiothiophene-2-carbaldehyde, 5-methylmercaptothiophene-2-carbaldehyde
PubChem CID 2736997
IUPAC Name 5-methylsulfanylthiophene-2-carbaldehyde
SMILES CSC1=CC=C(S1)C=O

Specifications

Quantity 1 g
Formula Weight 158.24
Percent Purity 97%
Chemical Name or Material 5-(Methylthio)thiophene-2-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.