missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrobenzene-1,3-Dicarboxylic Acid, TRC

Product Code. 17984755
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17984755 10 g 10g
17940445 1 g 1g
17991915 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17984755 Supplier Toronto Research Chemicals Supplier No. TRCN49479010G
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-88-2
Molecular Formula C8 H5 N O6
Molecular Weight (g/mol) 211.13
Synonym 5-Nitroisophtalic Acid, Glycopyrrolate Related Compound A (USP), 5-Nitrobenzene-1,3-dicarboxylic acid, 5-Nitro-isophthalic acid
IUPAC Name 5-nitrobenzene-1,3-dicarboxylic acid
SMILES OC(=O)c1cc(cc(c1)[N+](=O)[O-])C(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H5NO6/c10-7(11)4-1-5(8(12)13)3-6(2-4)9(14)15/h1-3H,(H,10,11)(H,12,13)
Formula Weight 211.0117
Chemical Name or Material 5-Nitro-isophthalic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.