missing translation for 'onlineSavingsMsg'
Learn More

5-Nitroisophthalic acid, 97+%

Product Code. 11479203
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479203 100 g 100g
11489203 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479203 Supplier Thermo Scientific Alfa Aesar Supplier No. A11696.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 618-88-2 | C8H5NO6 | 211.129 g/mol

5-Nitroisophthalic acid is used as an intermediate of medicine compound and as a disperser in dyes. It is used as an intermediate in organic synthesis,agrochemical,pharmaceutical and dyestuffs etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitroisophthalic acid is used as an intermediate of medicine compound and as a disperser in dyes. It is used as an intermediate in organic synthesis,agrochemical,pharmaceutical and dyestuffs etc,.

Solubility
Soluble in water.1.5 g/L at 20°C

Notes
Incompatible with strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-88-2
Molecular Formula C8H5NO6
Molecular Weight (g/mol) 211.129
MDL Number MFCD00007254
InChI Key NBDAHKQJXVLAID-UHFFFAOYSA-N
Synonym 5-nitroisophthalic acid, 5-nitrobenzene-1,3-dicarboxylic acid, 5-nitro-1,3-benzenedicarboxylic acid, 1,3-benzenedicarboxylic acid, 5-nitro, 5-nitroisophthalicacid, 5-nitro-m-phthalic acid, unii-g7rz7lc76u, isophthalic acid, 5-nitro, 1-nitrobenzene-3,5-dicarboxylic acid, g7rz7lc76u
PubChem CID 12069
SMILES C1=C(C=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 260°C to 264°C
Odor Odorless
Quantity 100 g
Beilstein 1976587
Solubility Information Soluble in water.1.5g/L at 20°C
Formula Weight 211.13
Percent Purity 98%
Chemical Name or Material 5-Nitroisophthalic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.