missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrosalicylic acid, 99%

Product Code. 10309070 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
250g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10309070 25 g Plastic bottle 25g
10237320 5 g Glass Bottle 5g
10703151 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10309070 Supplier Thermo Scientific Supplier No. 300480250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-97-9 | C7H5NO5 | 183.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-97-9
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.12
MDL Number MFCD00007338
InChI Key PPDRLQLKHRZIJC-UHFFFAOYSA-N
Synonym 5-nitrosalicylic acid, 2-hydroxy-5-nitrobenzoic acid, anilotic acid, benzoic acid, 2-hydroxy-5-nitro, 5-nitro-2-hydroxybenzoic acid, unii-82l9g7fyz3, 2-hydroxy-5-nitro-benzoic acid, 5-nitro-salicylic acid, salicylic acid, 5-nitro, 5-nitro salicylic acid
PubChem CID 7318
ChEBI CHEBI:61281
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 228°C to 231°C
Color Beige-Yellow to Brown-Orange
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula O2NC6H3(OH)COOH
Quantity 25 g
Beilstein 10, 116
Merck Index 15, 6718
Solubility Information Solubility in water: 1g/1475mL in water. Other solubilities: more soluble in hot water,very soluble in ethanol,ether,acetone
Formula Weight 183.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 5-Nitrosalicylic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Nitrosalicylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 5-Nitrosalicylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.