missing translation for 'onlineSavingsMsg'
Learn More

5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-2'-deoxyuridine, 98%

Product Code. 15480347
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15480347 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15480347 Supplier Thermo Scientific Alfa Aesar Supplier No. J64217.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 146954-74-7 | C30H29FN2O7 | 548.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 146954-74-7
Color White to Yellow
Molecular Formula C30H29FN2O7
MDL Number MFCD06657648
Quantity 5 g
Synonym 1-2r,3r,4r,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-3-fluoro-4-hydroxytetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, 5'-o-bis 4-methoxyphenyl phenylmethyl-2'-deoxy-2'-fluoro-uridine, 2'-deoxy-5'-o-dmt-2'-fluoro-uridine, 2'-deoxy-5'-o-dmt-2'-fluoro-d-uridine, 5'-o-bis 4-methoxyphenyl phenyl methyl-2'-deoxy-2'-fluorouridine, 5'-o-dmt-2'-fluoro-2'-deoxyuridine, 5'-o-4,4'-dimethoxytrityl-2'-fluoro-2'-deoxyuridine, uridine, 5'-o-bis 4-methoxyphenyl phenylmethyl-2'-deoxy-2'-fluoro, 1-2r,3r,4r,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-3-fluoro-4-hydroxyoxolan-2-yl-3h-pyrimidine-2,4-dione, 2'-fluoro-5'-o-dmt-uridine
InChI Key CSSFZSSZXOCCJB-QSZUMSSQNA-N
SMILES COC1=CC=C(C=C1)C(OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=CC(=O)NC1=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1
IUPAC Name 1-[(2R,3R,4R,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-3-fluoro-4-hydroxyoxolan-2-yl]pyrimidine-2,4-dione
Molecular Weight (g/mol) 548.57
PubChem CID 7073186
Formula Weight 548.56
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-2'-deoxyuridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.