missing translation for 'onlineSavingsMsg'
Learn More

5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-methylguanosine, 98%

Product Code. 15440457
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15440457 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15440457 Supplier Thermo Scientific Alfa Aesar Supplier No. J65583.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 114745-26-5 | C36H39N5O8 | 669.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in acetonitrile

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Specifications

CAS 114745-26-5
Color White to Yellow
Molecular Formula C36H39N5O8
MDL Number MFCD09842119
Quantity 1 g
Synonym 5'-o-dmt-n2-isobutyryl-2'-o-methyl-d-guanosine, n-9-2r,3r,4r,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-4-hydroxy-3-methoxytetrahydrofuran-2-yl-6-oxo-6,9-dihydro-1h-purin-2-yl isobutyramide, 5'-o-dmt-n2-isobutyryl-2'-o-methyl-guanosine, n-9-2r,3r,4r,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-4-hydroxy-3-methoxyoxolan-2-yl-6-oxo-1h-purin-2-yl-2-methylpropanamide, 5-o-dmt-n2-isobutyryl-2-o-methyl-guanosine, 5'-o-4,4'-dimethoxytrityl-n2-isobutyryl-2'-o-methylguanosine, 5'-o-bis 4-methoxyphenyl phenyl methyl-2'-o-methyl-n-2-methylpropanoyl guanosine, guanosine,5'-o-bis 4-methoxyphenyl phenylmethyl-2'-o-methyl-n-2-methyl-1-oxopropyl
Solubility Information Soluble in acetonitrile
InChI Key ISQLJOGRNUQHJX-QTXIGGTANA-N
SMILES CO[C@@H]1[C@H](O)[C@@H](COC(C2=CC=CC=C2)(C2=CC=C(OC)C=C2)C2=CC=C(OC)C=C2)O[C@H]1N1C=NC2=C1NC(NC(=O)C(C)C)=NC2=O
IUPAC Name N-[9-[(2R,3R,4R,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-4-hydroxy-3-methoxyoxolan-2-yl]-6-oxo-3H-purin-2-yl]-2-methylpropanamide
Molecular Weight (g/mol) 669.74
PubChem CID 14991812
Formula Weight 669.72
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-methylguanosine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.