missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-2,4-pentadienoic acid, 98+%

Product Code. 11354414
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11354414 25 g 25g
11344414 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354414 Supplier Thermo Scientific Alfa Aesar Supplier No. L01004.14
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1552-94-9 | C11H10O2 | 174.199 g/mol

5-Phenyl-2,4-pentadienoic acid is used as Synthetic building block, anti-malarial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Phenyl-2,4-pentadienoic acid is used as Synthetic building block, anti-malarial agent.

Solubility
Insoluble in water.

Notes
Storage temperature: 2 -4°C and Keep container tightlyclosed in a dryand well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1552-94-9
Molecular Formula C11H10O2
Molecular Weight (g/mol) 174.199
MDL Number MFCD00014018
InChI Key FEIQOMCWGDNMHM-KBXRYBNXSA-N
Synonym 5-phenylpenta-2,4-dienoic acid, 2e,4e-5-phenylpenta-2,4-dienoic acid, 5-phenyl-2,4-pentadienoic acid, 2,4-pentadienoic acid, 5-phenyl, e,e-cinnamylideneacetic acid, 2e,4e-5-phenyl-2,4-pentadienoic acid, 2,4-pentadienoicacid, 5-phenyl-, 2e,4e, 4-phenyl-1,3-butadiene-1-carboxylic acid, juarezic acid, cinnamylidene acetic acid
PubChem CID 1549512
IUPAC Name (2E,4E)-5-phenylpenta-2,4-dienoic acid
SMILES C1=CC=C(C=C1)C=CC=CC(=O)O

Specifications

Melting Point 163°C to 167°C
Quantity 25 g
Beilstein 2042414
Solubility Information Insoluble in water.
Formula Weight 174.2
Percent Purity ≥98%
Chemical Name or Material 5-Phenyl-2,4-pentadienoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.