missing translation for 'onlineSavingsMsg'
Learn More

5-R-Rivaroxaban, MedChemExpress

Product Code. 16498218
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498218 10 mg 10mg
16490658 5 mg 5mg
16454117 50 mg 50mg
16447956 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498218 Supplier Medchem Express Supplier No. HY76948
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 5-R-Rivaroxaban is (R)-enantiomer of Rivaroxaban. Rivaroxaban (BAY 59-7939) is a highly potent and selective, direct Factor Xa (FXa) inhibitor, achieving a strong gain in anti-FXa potency (IC50 0.7 nM; Ki 0.4 nM).

5-R-Rivaroxaban is (R)-enantiomer of Rivaroxaban. Rivaroxaban (BAY 59-7939) is a highly potent and selective, direct Factor Xa (FXa) inhibitor, achieving a strong gain in anti-FXa potency (IC50 0.7 nM

Specifications

Chemical Name or Material 5-R-Rivaroxaban
Physical Form Solid
CAS 865479-71-6
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C19H18ClN3O5S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 33.33 mg/mL (76.47 mM; Need ultrasonic)
SMILES O=C(O[C@@H]1CNC(C2=CC=C(Cl)S2)=O)N(C1)C(C=C3)=CC=C3N4C(COCC4)=O
Molecular Weight (g/mol) 435.88
Formula Weight 435.88
Percent Purity 99.72%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.