missing translation for 'onlineSavingsMsg'
Learn More

5-Sulfosalicylic acid dihydrate, 98%

Product Code. 11475533
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475533 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11475533 Supplier Thermo Scientific Alfa Aesar Supplier No. A13461.18
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5965-83-3 | C9H14O6S | 250.27 g/mol

5-Sulfosalicylic acid dihydrate may be employed for the quantification of doxorubicin derived from PEGylated liposomal doxorubicin (Doxil) and its major metabolite in human plasma by HPLC. It may be used in the preparation of poly[[tris(di-2-pyridylamine)tris([?]-5-sulfonatosalicylato)tricopper(II)] trihydrate]. Also used as an intermediate synthesis of bulk drugs, surfactant and additive for lubricating grease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Sulfosalicylic acid dihydrate may be employed for the quantification of doxorubicin derived from PEGylated liposomal doxorubicin (Doxil) and its major metabolite in human plasma by HPLC. It may be used in the preparation of poly[[tris(di-2-pyridylamine)tris([μ]-5-sulfonatosalicylato)tricopper(II)] trihydrate]. Also used as an intermediate synthesis of bulk drugs, surfactant and additive for lubricating grease.

Solubility
Soluble in water

Notes
Light Sensitive, store in dark. Store in cool, dry place in tightly closed container. Store away from strong bases and oxidizing agent.
TRUSTED_SUSTAINABILITY

Specifications

CAS 5965-83-3
Melting Point 103°C to 112°C
Assay Percent Range 99%
Odor Pungent
Molecular Formula C9H14O6S
MDL Number MFCD00007508
Quantity 50 g
UN Number UN2585
Beilstein 650741
Sensitivity Light sensitive
Merck Index 14,8964
Synonym 2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09 ngq462s6
Solubility Information Soluble aq. soln.
InChI Key NFYHZVWMQHQKRU-UHFFFAOYSA-N
SMILES C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
IUPAC Name 2-hydroxy-5-sulfobenzoic acid;dihydrate
Molecular Weight (g/mol) 250.27
PubChem CID 2723734
Formula Weight 254.22 (218.19 Anhydrous)
Percent Purity 98%
Chemical Name or Material 5-Sulfosalicylic acid dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.