missing translation for 'onlineSavingsMsg'
Learn More

5-Sulfosalicylic acid dihydrate, ACS, 99+%

Product Code. p-7058734
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11354818

Brand: Thermo Scientific Alfa Aesar 043144.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reagent for precipitation of proteins | CAS: 5965-83-3 | C9H14O6S | 250.27 g/mol

Reagent for precipitation of proteins5-Sulfosalicylic acid dihydrate is used as a metal scavenger and it forms proton-transfer dye complexes with diazo compounds such as 4-(phenyldiazenyl)aniline. It is employed as a preparative measure for removing proteins prior to chromatographic analysis. It plays a vital role in reducing and fixation of proteins in agarose and polyacrylamide gels. It acts as a surfactant and lubricating grease additive. It is also used in urine tests to determine urine protein content. Further, it acts as an intermediate in the synthesis of active pharmaceutical ingredients like doxycycline and methacycline. In addition to this, it is used as a spray reagent for the detection of amino acids on thin layer chromatography (TLC) plates and as a doping agent for polyaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5965-83-3
Molecular Formula C9H14O6S
Molecular Weight (g/mol) 250.27
MDL Number MFCD00007508,MFCD00149540
InChI Key NFYHZVWMQHQKRU-UHFFFAOYSA-N
Synonym 2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
PubChem CID 2723734
IUPAC Name 2-hydroxy-5-sulfobenzoic acid;dihydrate
SMILES C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O

Specifications

Melting Point 103°C to 112°C
Quantity 25 g
UN Number UN2585
Beilstein 650741
Sensitivity Light sensitive
Merck Index 14,8964
Solubility Information Soluble in water,alcohol,ether and polar solvents.
Formula Weight 254.22 (218.19 Anhydrous)
Percent Purity ≥99%
Grade ACS
Physical Form Powder
Chemical Name or Material 5-Sulfosalicylic Acid, Dihydrate
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 5-Sulfosalicylic acid dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.