missing translation for 'onlineSavingsMsg'
Learn More

Dabrafenib Mesylate, MedChemExpress

Product Code. 16461746
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16461746

Brand: Medchem Express HY14660A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dabrafenib Mesylate is a potent and selective Raf kinase inhibitor with IC50s of 0.6 and 5.0 nM for RafV600E and c-Raf, respectively.

Dabrafenib Mesylate is a potent and selective Raf kinase inhibitor with IC50s of 0.6 and 5.0 nM for RafV600E and c-Raf, respectively.

Specifications

Chemical Name or Material Dabrafenib Mesylate
CAS 1195768-06-9
Color White
Purity Grade Notes Research
Molecular Formula C24H24F3N5O5S3
Quantity 500 mg
Synonym GSK2118436 Mesylate GSK 2118436B
Solubility Information DMSO : ≥ 36 mg/mL (58.47 mM)
SMILES CC(C)(C)C1=NC(C2=C(F)C(NS(C3=C(F)C=CC=C3F)(=O)=O)=CC=C2)=C(C4=CC=NC(N)=N4)S1.CS(=O)(O)=O
Molecular Weight (g/mol) 615.67
Formula Weight 615.67
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.