missing translation for 'onlineSavingsMsg'
Learn More

Cefditoren (Pivoxil), MedChemExpress

Product Code. 16483367
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16483367

Brand: Medchem Express HY17452A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cefditoren Pivoxil (ME 1207) is a broad-spectrum, third-generation, oral cephalosporin antibacterial with enhanced stability against many common β lactamases.

Cefditoren Pivoxil has activity against Gram-negative organisms and Gram-positive organisms. Cefditoren Pivoxil can be used in the research of infection diseases such as acute exacerbations of chronic bronchitis, community-acquired pneumonia (CAP), streptococcal pharyngitis/tonsillitis, or uncomplicated skin and skin structure infections.

Specifications

Chemical Name or Material Cefditoren (Pivoxil)
Physical Form Solid
CAS 117467-28-4
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C25H28N6O7S3
For Use With (Application) COVID-19-immunoregulation
Synonym Cefditoren pivoxyl Cefditoren pivaloyloxymethyl ester ME 1207
Solubility Information DMSO : ≥ 100 mg/mL (161.10 mM)
SMILES O=C(C(N12)=C(/C=C\C3=C(C)N=CS3)CS[C@]2([H])[C@H](NC(/C(C4=CSC(N)=N4)=N\OC)=O)C1=O)OCOC(C(C)(C)C)=O
Molecular Weight (g/mol) 620.72
Formula Weight 620.72
Percent Purity 99.31%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.