missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso-Salbutamol, TRC

Product Code. 17901385
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17901385 50 mg 50mg
17922035 10 mg 10mg
17954755 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17901385 Supplier Toronto Research Chemicals Supplier No. TRCN51450050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2919946-71-5
Molecular Formula C13 H20 N2 O4
Molecular Weight (g/mol) 268.31
Synonym N-(tert-Butyl)-N-(2-hydroxy-2-(4-hydroxy-3-(hydroxymethyl)phenyl)ethyl)nitrous Amide, N-Nitrososalbutamol, N-Tert-butyl-N-[2-hydroxy-2-[4-hydroxy-3-(hydroxymethyl)phenyl]ethyl]nitrous amide, N-Nitrososalbutamol
IUPAC Name N-tert-butyl-N-[2-hydroxy-2-[4-hydroxy-3-(hydroxymethyl)phenyl]ethyl]nitrous amide
SMILES CC(C)(C)N(CC(O)c1ccc(O)c(CO)c1)N=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H20N2O4/c1-13(2,3)15(14-19)7-12(18)9-4-5-11(17)10(6-9)8-16/h4-6,12,16-18H,7-8H2,1-3H3
Formula Weight 268.1423
Percent Purity >95%
Chemical Name or Material N-Nitroso-Salbutamol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.