missing translation for 'onlineSavingsMsg'
Learn More

53BP1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-304SS

 View more versions of this product

Product Code. 18735403

  • ONLINE EXCLUSIVE PRICE
    315.00 €
    297.15 € / 25µg
    Save 17.85 €
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

53BP1 Polyclonal specifically detects 53BP1 in Human, Mouse, Rat, Bat, Bovine, Canine, Fish, Goat, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, In-situ Hybridization, Flow (Intracellular), Knockout Validated, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

53BP1
Polyclonal
Unconjugated
PBS with 0.05% Sodium Azide
53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1
Rabbit
214 kDa
0.025 mL
Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Tumor Suppressors
7158
Human, Mouse, Rat, Bat, Bovine, Canine, Fish, Goat, Primate
IgG
Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, In Situ Hybridization (ISH), Immunoassay, KnockDown
1 mg/mL
Western Blot 1:5000-1:25000, Chromatin Immunoprecipitation reported in scientific literature (PMID 24591601), Flow Cytometry 1.5 ug/ml, Immunohistochemistry reported in scientific literature (PMID 24987917), Immunocytochemistry/Immunofluorescence 1:1000-1:5000, Immunoprecipitation reported in scientific literature (PMID 25645366), Immunohistochemistry-Paraffin 1:1000-1:5000. Use reported in scientific literature (PMID 27653664), Immunohistochemistry-Frozen reported by customer review, Immunoblotting reported in multiple pieces of scientific literature, In-situ Hybridization reported in scientific literature (PMID 34988401; 33122290), Flow (Intracellular), Chromatin Immunoprecipitation (ChIP), Knockout Validated reported in scientific literature (PMID 26601238), KnockDown Validated
Q12888
TP53BP1
The epitope recognized by 53BP1 Antibody maps to a region between residues 350 and 400 of human 53BP1 [NP_005648.1].
Affinity Purified
RUO
Primary
Human, mouse, rat and goat. Human has been tested in WB, IHC-P and ICC/IF, mouse and goat have only been tested in ICC/IF. Primate reactivity reported in scientific literature (PMID: 18389475).Fish reactivity reported in scientific literature (PMID: 25516420).
Store at -20 degreesC.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only