missing translation for 'onlineSavingsMsg'
Learn More

53BP1 Antibody Pack, Novus Biologicals™

Product Code. 18551972 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Pack
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18551972 1 Pack 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18551972 Supplier Novus Biologicals Supplier No. NB1009261Pack

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody Pack

53BP1 Antibody Packantibody specifically detects 53BP1 in Human, Mouse, Rat, Bat, Bovine, Canine, Fish, Goat, Naked mole-rat, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, In-situ Hybridization, Flow (Intracellular), ChIP assay (ChIP), Gene Knock-Out, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen 53BP1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation Tris-citrate/phosphate buffer, pH 7 to 8
Gene Alias 53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1
Immunogen See individual datasheets.
Purification Method Immunogen affinity purified
Quantity 1 Pack
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Tumor Suppressors
Gene ID (Entrez) 7158
Target Species Human, Mouse, Rat, Bat, Bovine
Content And Storage Store at -20°C.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.