missing translation for 'onlineSavingsMsg'
Learn More

Δ5a-11-Hydroxy-12-oxo Minocycline Disodium Salt, TRC

Product Code. 30406619
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
5 mg
Unit Size:
0.5mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30406619 0.5 mg 0.5mg
30396731 5 mg 5mg
30394660 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406619 Supplier Toronto Research Chemicals Supplier No. TRCH9489250.5MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Delta5a-11-Hydroxy-12-oxo Minocycline Disodium Salt
Molecular Formula C23 H23 N3 O7 . 2Na
Quantity 0.5 mg
Synonym Δ5a-11-Hydroxy-12-oxo Minocycline Disodium Salt, 4,7-Bis(dimethylamino)-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-1,12-dioxo-2-naphthacenecarboxamide Disodium Salt, 5a,6-Anhydro Minocycline Disodium Salt
InChI Formula InChI=1S/C23H25N3O7.2Na/c1-25(2)12-5-6-13(27)15-10(12)7-9-8-11-17(26(3)4)19(29)16(22(24)32)21(31)23(11,33)20(30)14(9)18(15)28;;/h5-7,11,17,27-29,33H,8H2,1-4H3,(H2,24,32);;/q;2*+1/p-2/t11-,17-,23-;;/m0../s1
SMILES [Na+].[Na+].CN(C)[C@H]1[C@@H]2Cc3cc4c(ccc([O-])c4c(O)c3C(=O)[C@]2(O)C(=O)C(=C1[O-])C(=O)N)N(C)C
IUPAC Name disodium;(6aS,7S,10aS)-9-carbamoyl-4,7-bis(dimethylamino)-10a,12-dihydroxy-10,11-dioxo-6a,7-dihydro-6H-tetracene-1,8-diolate
Molecular Weight (g/mol) 499.42
Formula Weight 499.1331
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.