missing translation for 'onlineSavingsMsg'
Learn More

5H-Perfluoropentanoic acid, 96%

Product Code. p-7046957
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11353856

Brand: Thermo Scientific Alfa Aesar L16960.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 376-72-7 | C5H2F8O2 | 246.056 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 376-72-7
Molecular Formula C5H2F8O2
Molecular Weight (g/mol) 246.056
MDL Number MFCD00054690
InChI Key VGFKXVSMDOKOJZ-UHFFFAOYSA-N
Synonym 5h-octafluoropentanoic acid, octafluoropentanoic acid, 2,2,3,3,4,4,5,5-octafluorovaleric acid, 5h-perfluoropentanoic acid, omega-h-oktafluorpentansaeure german, pentanoic acid, 2,2,3,3,4,4,5,5-octafluoro, omega-h-oktafluorpentansaeure, 5-h-octafluoropentanoic acid, 4-02-00-00877 beilstein handbook reference
PubChem CID 120227
IUPAC Name 2,2,3,3,4,4,5,5-octafluoropentanoic acid
SMILES C(C(C(C(C(=O)O)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point 0°C
Density 1.725
Boiling Point 165°C to 166°C
Refractive Index 1.319
Quantity 5 g
UN Number UN3265
Beilstein 1797233
Formula Weight 246.06
Percent Purity 96%
Chemical Name or Material 5H-Perfluoropentanoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.