missing translation for 'onlineSavingsMsg'
Learn More

KN-92 phosphate, MedChemExpress

Product Code. 16409167
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16409167 5 mg 5mg
16450246 2 mg 2mg
16432687 50 mg 50mg
16406598 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16409167 Supplier Medchem Express Supplier No. HY15517A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KN-92 phosphate is an inactive derivative of KN-93, without CaM kinase inhibitory activity. KN-92 phosphate is intended to be used as a control compound in studies designed to elucidate the antagonist activities of KN-93. KN-93 is a cell-permeable, reversible and competitive CaMKII inhibitor.

KN-92 phosphate is an inactive derivative of KN-93, without CaM kinase inhibitory activity. KN-92 phosphate is intended to be used as a control compound in studies designed to elucidate the antagonist activities of KN-93. KN-93 is a cell-permeable, reversible and competitive CaMKII inhibitor.

Specifications

Chemical Name or Material KN-92 phosphate
CAS 1135280-28-2
Color White
Purity Grade Notes Research
Molecular Formula C24H28ClN2O7PS
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (180.19 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=S(C1=CC=C(OC)C=C1)(NC2=CC=CC=C2CN(C/C=C/C3=CC=C(Cl)C=C3)C)=O.O=P(O)(O)O
Molecular Weight (g/mol) 554.98
Formula Weight 554.98
Percent Purity 99.35%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.