missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(RS)-MCPG, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
(RS)-MCPG (alpha-MCPG) is a competitive and selective group I/group II metabotropic glutamate receptor (mGluR) antagonist. (RS)-MCPG blocks theta-burst stimulation (TBS)-induced shifts in both juvenile and neonatal rat hippocampal neurons.
Specifications
Specifications
| Chemical Name or Material | (RS)-MCPG |
| Physical Form | Powder |
| CAS | 146669-29-6 |
| Color | Off-White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C10H11NO4 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Synonym | alpha-MCPG |
| Solubility Information | 1M NaOH : 100 mg/mL (478.01 mM; ultrasonic and adjust pH to 10 with NaOH) ∣DMSO : 6 mg/mL (28.68 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (ultrasonic;heat to 80°C) (insoluble) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction