missing translation for 'onlineSavingsMsg'
Learn More

6,12-Dibromochrysene, 99%, Thermo Scientific™

Product Code. 15436848
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436848 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15436848 Supplier Thermo Scientific Alfa Aesar Supplier No. H60383.MF
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 131222-99-6
Molecular Formula C18H10Br2
MDL Number MFCD00092281
Quantity 50 mg
Synonym 6,12-dibromochryscene, chrysene, 6,12-ibromo, chrysene, 6,12-dibromo, acmc-1c5x4, 6,12-bis bromanyl chrysene, 6,12-dibromochrysen
InChI Key RULVBMDEPWAFIN-UHFFFAOYSA-N
SMILES BrC1=CC2=C(C=C(Br)C3=CC=CC=C23)C2=CC=CC=C12
IUPAC Name 6,12-dibromochrysene
Molecular Weight (g/mol) 386.09
PubChem CID 3286225
Formula Weight 386.088
Percent Purity 99%
Chemical Name or Material 6,12-Dibromochrysene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.