missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dichloro-1-tetralone, TRC

Product Code. 30377113
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30377113 100 mg
30376495 500 mg
30368962 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30377113 Supplier Toronto Research Chemicals Supplier No. TRCD439878100MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25095-57-2
Molecular Formula C10H8OCl2
Molecular Weight (g/mol) 215.08
SMILES Clc1cc2CCCC(=O)c2cc1Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H8Cl2O/c11-8-4-6-2-1-3-10(13)7(6)5-9(8)12/h4-5H,1-3H2
Formula Weight 213.9952
Chemical Name or Material 6,7-Dichloro-1-tetralone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.