missing translation for 'onlineSavingsMsg'
Learn More

6,8-Dihydro-7H-pyrrolo[2,3-g]benzothiazol-7-one, TRC

Product Code. 30373589
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30373589 50 mg
30377179 25 mg
30383001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30373589 Supplier Toronto Research Chemicals Supplier No. TRCD45365850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 222036-27-3
Molecular Formula C9H6N2OS
Molecular Weight (g/mol) 190.22
Synonym 6H-Thiazolo[5,4-e]indol-7(8H)-one
SMILES O=C1Cc2c(N1)ccc3ncsc23

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H6N2OS/c12-8-3-5-6(11-8)1-2-7-9(5)13-4-10-7/h1-2,4H,3H2,(H,11,12)
Chemical Name or Material 6,8-Dihydro-7H-pyrrolo[2,3-g]benzothiazol-7-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.