missing translation for 'onlineSavingsMsg'
Learn More

6-Acetyl-1,2,3,4-tetrahydronaphthalene, 97%

Product Code. 11467636
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467636 5 g 5g
11477636 25 g 25g
11487636 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467636 Supplier Thermo Scientific Alfa Aesar Supplier No. A14666.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 774-55-0 | C12H14O | 174.24 g/mol

6-Acetyl-1,2,3,4-tetrahydronaphthalene, is used asa novel methyl anthranilate yet extremely stable and resistant to discoloration. It is used in liquid detergent, concentrated fabric softner, alcoholic fine fragrances, antiperspirants. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Acetyl-1,2,3,4-tetrahydronaphthalene, is used asa novel methyl anthranilate yet extremely stable and resistant to discoloration. It is used in liquid detergent, concentrated fabric softner, alcoholic fine fragrances, antiperspirants. It is also used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Product is sensitive to light and moisture. Handle and store under inert gas. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 774-55-0
Molecular Formula C12H14O
Molecular Weight (g/mol) 174.24
MDL Number MFCD00019710
InChI Key VEPUKHYQNXSSKV-UHFFFAOYSA-N
Synonym 1-5,6,7,8-tetrahydronaphthalen-2-yl ethanone, 6-acetyltetralin, 6-acetyl tetralin, ethanone, 1-5,6,7,8-tetrahydro-2-naphthalenyl, unii-6l3dbr6de6, 6-acetyl-1,2,3,4-tetrahydronaphthalene, 5,6,7,8-tetrahydro-2-acetonaphthone, 6l3dbr6de6, 6-acetyltetrahydronaphthalene, 1-5,6,7,8-tetrahydronaphthalen-2-yl ethan-1-one
PubChem CID 69885
SMILES CC(=O)C1=CC=C2CCCCC2=C1

Specifications

Melting Point -21°C
Density 1.058
Boiling Point 119°C to 121°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.561
Quantity 5 g
Beilstein 1241458
Solubility Information Slightly soluble in water.
IUPAC Name 1-(5,6,7,8-tetrahydronaphthalen-2-yl)ethanone
Formula Weight 174.24
Percent Purity 97%
Chemical Name or Material 6-Acetyl-1,2,3,4-tetrahydronaphthalene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.