missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-1-tetralone, 97%

Product Code. 15464697
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15464697 10 g 10g
15454697 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15464697 Supplier Thermo Scientific Alfa Aesar Supplier No. H32028.09
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3470-53-9 | C10H11NO | 161.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3470-53-9
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD00099462
InChI Key BEVVUJBVEXJGKM-UHFFFAOYSA-N
Synonym 6-amino-1,2,3,4-tetrahydronaphthalen-1-one, 6-amino-1-tetralone, 6-amino-3,4-dihydronaphthalen-1 2h-one, 6-amino-3,4-dihydro-1 2h-naphthalenone, 1 2h-naphthalenone, 6-amino-3,4-dihydro, 6-amino-1,2,3,4-tetrahydro napthalene-1-one, 6-amino-2,3,4-trihydronaphthalen-1-one, 6-azanyl-3,4-dihydro-2∼ h-naphthalen-1-one, 6-aminotetralone, pubchem12409
PubChem CID 339537
IUPAC Name 6-amino-3,4-dihydro-2H-naphthalen-1-one
SMILES C1CC2=C(C=CC(=C2)N)C(=O)C1

Specifications

Melting Point 128°C to 130°C
Quantity 10 g
UN Number UN2811
Sensitivity Moisture sensitive
Formula Weight 161.21
Percent Purity 97%
Chemical Name or Material 6-Amino-1-tetralone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.