missing translation for 'onlineSavingsMsg'
Learn More

6-Aminocaproic acid, MedChemExpress

Product Code. 16438186
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438186 100 mg 100mg
16404317 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16438186 Supplier Medchem Express Supplier No. HYB0236
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 6-Aminocaproic acid (EACA), a monoamino carboxylic acid, is a potent and orally active inhibitor of plasmin and plasminogen. 6-Aminocaproic acid is a potent antifibrinolytic agent. 6-Aminocaproic acid prevents clot lysis through the competitive binding of lysine residues on plasminogen, inhibiting plasmin formation and reducing fibrinolysis. 6-Aminocaproic acid can be used for the research of bleeding disorders.

6-Aminocaproic acid (EACA), a monoamino carboxylic acid, is a potent and orally active inhibitor of plasmin and plasminogen. 6-Aminocaproic acid is a potent antifibrinolytic agent. 6-Aminocaproic acid prevents clot lysis through the competitive binding of lysine residues on plasminogen, inhibiting plasmin formation and reducing fibrinolysis. 6-Aminocaproic acid can be used for the research of bleeding disorders.

Specifications

Chemical Name or Material 6-Aminocaproic acid
Physical Form Solid
CAS 60-32-2
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C6H13NO2
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym EACA Epsilon-Amino-n-caproic Acid 6-Aminohexanoic acid
Solubility Information H2O : ≥ 50 mg/mL (381.18 mM) ∣DMSO : 1 mg/mL (7.62 mM; Need ultrasonic)
SMILES O=C(O)CCCCCN
Molecular Weight (g/mol) 131.17
Formula Weight 131.17
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.