missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Aminocoumarin hydrochloride, 97%
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Beschreibung
6-Aminocoumarin hydrochloride is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Aminocoumarin hydrochloride is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
6-Aminocoumarin hydrochloride is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
Spezifikation
Spezifikation
| Melting Point | >260°C (decomposition) |
| Quantity | 1 g |
| Beilstein | 3698955 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 197.62 |
| Percent Purity | 97% |
| Chemical Name or Material | 6-Aminocoumarin hydrochloride |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu