missing translation for 'onlineSavingsMsg'
Learn More

6-Aminocoumarin hydrochloride, 97%

Product Code. 11397606
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397606 5 g 5g
11387606 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397606 Supplier Thermo Scientific Alfa Aesar Supplier No. L11168.06
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 63989-79-7 | C9H8ClNO2 | 197.62 g/mol

6-Aminocoumarin hydrochloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Aminocoumarin hydrochloride is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63989-79-7
Molecular Formula C9H8ClNO2
Molecular Weight (g/mol) 197.62
MDL Number MFCD00082671
InChI Key OSIGAIXSSYAHEG-UHFFFAOYSA-N
Synonym 6-aminocoumarin hydrochloride, 6-amino-2h-chromen-2-one hydrochloride, 6-aminochromen-2-one hydrochloride, 2-oxochromen-6-yl azanium chloride, 6-aminocoumarin hci, acmc-20am07, 6-amino-chromen-2-one hydrochloride, 2-oxochromen-6-yl ammonium chloride, 6-amino-2h-chromen-2-one hydrochloride 1:1
PubChem CID 356789
IUPAC Name 6-aminochromen-2-one;hydrochloride
SMILES [Cl-].[NH3+]C1=CC=C2OC(=O)C=CC2=C1

Specifications

Melting Point >260°C (decomposition)
Quantity 5 g
Beilstein 3698955
Solubility Information Insoluble in water.
Formula Weight 197.62
Percent Purity 97%
Chemical Name or Material 6-Aminocoumarin hydrochloride
compliance-icons
Product Identifier
  • 6-Aminocoumarin hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 6-Aminocoumarin hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.