missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene, 97%, Thermo Scientific™

Product Code. 15467765
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467765 10 g 10g
15457765 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467765 Supplier Thermo Scientific Alfa Aesar Supplier No. H30436.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene is used in Synthesis and evaluation of synthetic retinoid derivatives as inducers of stem cell differentiation.

Solubility
Insoluble in water

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27452-17-1
Molecular Formula C14H19Br
Molecular Weight (g/mol) 267.21
MDL Number MFCD05664407
InChI Key NLOOVMVNNNYLFS-UHFFFAOYSA-N
Synonym 6-bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene, naphthalene, 6-bromo-1,2,3,4-tetrahydro-1,1,4,4-tetramethyl, 1,1,4,4-tetramethyl-6-bromotetralin, 6-bromo-1,1,4,4-tetramethyl-tetralin, 6-bromo-1,1,4,4 tetramethy 1,2,3,4-tetrahydronaphthalene, 6-bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphtalene, 2-bromo-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalene, 2-bromo-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaph-thalene, 2-bromo-5,6,7,8-tetrahydro-5,5,8,8-tetramethylnaphthalene
PubChem CID 226685
IUPAC Name 6-bromo-1,1,4,4-tetramethyl-2,3-dihydronaphthalene
SMILES CC1(CCC(C2=C1C=CC(=C2)Br)(C)C)C

Specifications

Melting Point 34°C to 37°C
Quantity 10 g
Solubility Information Insoluble in water
Formula Weight 267.21
Percent Purity 97%
Chemical Name or Material 6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.