missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene, Tech., Thermo Scientific™

Product Code. 10391533
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10391533 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10391533 Supplier Thermo Scientific Maybridge Supplier No. CC27110EA
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 27452-17-1
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Molecular Formula C14H19Br
MDL Number MFCD05664407
Quantity 10 g
Synonym 6-bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene, naphthalene, 6-bromo-1,2,3,4-tetrahydro-1,1,4,4-tetramethyl, 1,1,4,4-tetramethyl-6-bromotetralin, 6-bromo-1,1,4,4-tetramethyl-tetralin, 6-bromo-1,1,4,4 tetramethy 1,2,3,4-tetrahydronaphthalene, 6-bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphtalene, 2-bromo-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalene, 2-bromo-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaph-thalene, 2-bromo-5,6,7,8-tetrahydro-5,5,8,8-tetramethylnaphthalene
InChI Key NLOOVMVNNNYLFS-UHFFFAOYSA-N
SMILES CC1(CCC(C2=C1C=CC(=C2)Br)(C)C)C
IUPAC Name 6-bromo-1,1,4,4-tetramethyl-2,3-dihydronaphthalene
Molecular Weight (g/mol) 267.21
PubChem CID 226685
Formula Weight 267.21
Percent Purity 90%
Grade Technical
Chemical Name or Material 6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.