missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 6-Carboxyfluorescein, 96%

Product Code. 10285194 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10285194 100 mg Glass bottle 100mg
10634555 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10285194 Supplier Thermo Scientific Chemicals Supplier No. 425801000
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3301-79-9 | C21H12O7 | 376.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3301-79-9
Molecular Formula C21H12O7
Molecular Weight (g/mol) 376.32
MDL Number MFCD00036873
InChI Key BZTDTCNHAFUJOG-UHFFFAOYSA-N
Synonym 6-carboxyfluorescein, 6-fam, carboxyfluorescein, spiro isobenzofuran-1 3h ,9'-9h xanthene-6-carboxylic acid, 3',6'-dihydroxy-3-oxo, 3,6,9-trihydroxyxanthen-9-yl terephthalic acid, 3',6'-dihydroxy-3-oxo-3h-spiro 2-benzofuran-1,9'-xanthene-6-carboxylic acid, 3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-6-carboxylic acid, 5 6-carboxy fluorescein, bidd:gt0504
PubChem CID 76806
ChEBI CHEBI:39073
IUPAC Name 3',6'-dihydroxy-1-oxospiro[2-benzofuran-3,9'-xanthene]-5-carboxylic acid
SMILES C1=CC2=C(C=C1C(=O)O)C3(C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O)OC2=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point >300.0°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Formula Weight 376.32
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 6-Carboxyfluorescein
Show More Show Less
compliance-icons
Product Identifier
  • 6-Carboxyfluorescein
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.