missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 6-Carboxyfluorescein, 96%

Product Code. p-4089668 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 10285194

Brand: Thermo Scientific Chemicals 425801000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3301-79-9 | C21H12O7 | 376.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3301-79-9
Molecular Formula C21H12O7
Molecular Weight (g/mol) 376.32
MDL Number MFCD00036873
InChI Key BZTDTCNHAFUJOG-UHFFFAOYSA-N
Synonym 6-carboxyfluorescein, 6-fam, carboxyfluorescein, spiro isobenzofuran-1 3h ,9'-9h xanthene-6-carboxylic acid, 3',6'-dihydroxy-3-oxo, 3,6,9-trihydroxyxanthen-9-yl terephthalic acid, 3',6'-dihydroxy-3-oxo-3h-spiro 2-benzofuran-1,9'-xanthene-6-carboxylic acid, 3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-6-carboxylic acid, 5 6-carboxy fluorescein, bidd:gt0504
PubChem CID 76806
ChEBI CHEBI:39073
IUPAC Name 3',6'-dihydroxy-1-oxospiro[2-benzofuran-3,9'-xanthene]-5-carboxylic acid
SMILES C1=CC2=C(C=C1C(=O)O)C3(C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O)OC2=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point >300.0°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Formula Weight 376.32
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 6-Carboxyfluorescein
Show More Show Less
compliance-icons
Product Identifier
  • 6-Carboxyfluorescein
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.