missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-fluoropurine, 97%, Thermo Scientific Chemicals

Product Code. 11485540
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485540 500 mg 500mg
11475540 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11485540 Supplier Thermo Scientific Alfa Aesar Supplier No. H25825.ME
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1651-29-2 | Molecular Formula: C5H2ClFN4 | Molecular Weight: 172.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1651-29-2
Molecular Formula C5H2ClFN4
Molecular Weight (g/mol) 172.55
MDL Number MFCD02183557
InChI Key UNRIYCIDCQDGQE-UHFFFAOYSA-N
Synonym 6-chloro-2-fluoropurine, 6-chloro-2-fluoro-9h-purine, 2-fluoro-6-chloropurine, 6-chloro-2-fluoro-purine, cfp, 1jdj, pubchem9261, acmc-1c2uh, 2-fluoro,6-chloro-purine, 2-fluoro-6-chloro-purine
PubChem CID 5287914
IUPAC Name 6-chloro-2-fluoro-7H-purine
SMILES FC1=NC(Cl)=C2NC=NC2=N1

Specifications

Melting Point 151°C to 153°C
Quantity 500 mg
Formula Weight 172.55
Percent Purity 97%
Chemical Name or Material 6-Chloro-2-fluoropurine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.