missing translation for 'onlineSavingsMsg'
Learn More

6-Ethynylquinoxaline, ≥97%, Thermo Scientific™

Product Code. 10453295
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10453295 250 mg Amber Glass Bottle 250mg
10644575 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10453295 Supplier Thermo Scientific Maybridge Supplier No. CC11747CB
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 442517-33-1
Molecular Formula C10H6N2
Molecular Weight (g/mol) 154.172
MDL Number MFCD08435845
InChI Key QSYDWUWZZBZOCD-UHFFFAOYSA-N
PubChem CID 18525704
IUPAC Name 6-ethynylquinoxaline
SMILES C#CC1=CC2=NC=CN=C2C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym quinoxaline, 6-ethynyl, quinoxaline,6-ethynyl, quinoxalin-6-yl acetylene
Formula Weight 154.17
Percent Purity 97%
Chemical Name or Material 6-Ethynylquinoxaline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.