missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoroindole, 98%

Product Code. 11460498
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460498 25 g 25g
11440498 1 g 1g
11450498 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460498 Supplier Thermo Scientific Alfa Aesar Supplier No. B24633.14
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 399-51-9 | C8H6FN | 135.14 g/mol

6-Fluoroindole acts as a reagent in the synthesis of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles, which acts as a potential anticancer immunomodulator. It is also employed as an antifungal and antibacterial agent. Further, it serves as a potent selective serotonin reuptake inhibitor. In addition to this, it is used as a inhibitor of HIV-1 attachment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Fluoroindole acts as a reagent in the synthesis of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles, which acts as a potential anticancer immunomodulator. It is also employed as an antifungal and antibacterial agent. Further, it serves as a potent selective serotonin reuptake inhibitor. In addition to this, it is used as a inhibitor of HIV-1 attachment.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in a cool place.Incompatible with strong oxidizing agents, strong reducing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 399-51-9
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD00056933
InChI Key YYFFEPUCAKVRJX-UHFFFAOYSA-N
Synonym 6-fluoroindole, 1h-indole, 6-fluoro, 6-fluoro indole, 6-fluoroindol, 6-fluoro-indole, pubchem1682, acmc-1cty0, ksc222e4t, 6-fluoroindole 1g
PubChem CID 351278
IUPAC Name 6-fluoro-1H-indole
SMILES FC1=CC=C2C=CNC2=C1

Specifications

Melting Point 74°C to 77°C
Quantity 25 g
Beilstein 112192
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 135.14
Percent Purity 98%
Chemical Name or Material 6-Fluoroindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.