missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoroindole, 98%

Product Code. 10721175 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10721175 5 g Glass bottle 5g
10566071 250 mg Glass bottle 250mg
10123871 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10721175 Supplier Thermo Scientific Acros Supplier No. 342510050
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 399-51-9 | C8H6FN | 135.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 399-51-9
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD00056933
InChI Key YYFFEPUCAKVRJX-UHFFFAOYSA-N
Synonym 6-fluoroindole, 1h-indole, 6-fluoro, 6-fluoro indole, 6-fluoroindol, 6-fluoro-indole, pubchem1682, acmc-1cty0, ksc222e4t, 6-fluoroindole 1g
PubChem CID 351278
IUPAC Name 6-fluoro-1H-indole
SMILES FC1=CC=C2C=CNC2=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 72°C to 77°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 135.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 6-Fluoroindole
Show More Show Less
compliance-icons
Product Identifier
  • 6-Fluoroindole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.