missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-2-naphthoic acid, 99%

Product Code. 11430447
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430447 5 g 5g
11420447 1 g 1g
11440447 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430447 Supplier Thermo Scientific Alfa Aesar Supplier No. B21791.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16712-64-4 | C11H8O3 | 188.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16712-64-4
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.182
MDL Number MFCD00060070
InChI Key KAUQJMHLAFIZDU-UHFFFAOYSA-N
Synonym 6-hydroxy-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-hydroxy, 6-hydroxy-2-naphthalenecarboxylic acid, 6-carboxy-2-naphthol, 2-hydroxynaphthalene-6-carboxylic acid, 6-hydroxy-beta-naphthoic acid, unii-72xa858i6p, 6-hydroxy-naphthalene-2-carboxylic acid, 2-naphthoic acid, 6-hydroxy, 2-hydroxy-6-naphthoic acid
PubChem CID 85557
IUPAC Name 6-hydroxynaphthalene-2-carboxylic acid
SMILES C1=CC2=C(C=CC(=C2)O)C=C1C(=O)O

Specifications

Melting Point 248°C to 250°C
Quantity 5 g
Beilstein 2692084
Formula Weight 188.18
Percent Purity 99%
Chemical Name or Material 6-Hydroxy-2-naphthoic acid
compliance-icons
Product Identifier
  • 6-Hydroxy-2-naphthoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 6-Hydroxy-2-naphthoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.