missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-2-naphthoic acid, 99%

Product Code. 11440447
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440447 25 g 25g
11420447 1 g 1g
11430447 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440447 Supplier Thermo Scientific Alfa Aesar Supplier No. B21791.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16712-64-4 | C11H8O3 | 188.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 16712-64-4
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.182
MDL Number MFCD00060070
InChI Key KAUQJMHLAFIZDU-UHFFFAOYSA-N
Synonym 6-hydroxy-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-hydroxy, 6-hydroxy-2-naphthalenecarboxylic acid, 6-carboxy-2-naphthol, 2-hydroxynaphthalene-6-carboxylic acid, 6-hydroxy-beta-naphthoic acid, unii-72xa858i6p, 6-hydroxy-naphthalene-2-carboxylic acid, 2-naphthoic acid, 6-hydroxy, 2-hydroxy-6-naphthoic acid
PubChem CID 85557
IUPAC Name 6-hydroxynaphthalene-2-carboxylic acid
SMILES C1=CC2=C(C=CC(=C2)O)C=C1C(=O)O

Specifica

Melting Point 248°C to 250°C
Quantity 25 g
Beilstein 2692084
Formula Weight 188.18
Percent Purity 99%
Chemical Name or Material 6-Hydroxy-2-naphthoic acid

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.