missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy Warfarin, TRC

Product Code. 30133758
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133758 25 mg 25mg
17904665 5 mg 5mg
17971505 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133758 Supplier Toronto Research Chemicals Supplier No. TRCH99612025MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17834-02-5
Molecular Formula C19 H16 O5
Molecular Weight (g/mol) 324.33
Synonym 2H-1-Benzopyran-2-one, 4,6-dihydroxy-3-(3-oxo-1-phenylbutyl)-, Coumarin, 3-(α-acetonylbenzyl)-4,6-dihydroxy- (8CI), 6-Hydroxywarfarin
IUPAC Name 4,6-dihydroxy-3-(3-oxo-1-phenylbutyl)chromen-2-one
SMILES CC(=O)CC(C1=C(O)c2cc(O)ccc2OC1=O)c3ccccc3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H16O5/c1-11(20)9-14(12-5-3-2-4-6-12)17-18(22)15-10-13(21)7-8-16(15)24-19(17)23/h2-8,10,14,21-22H,9H2,1H3
Formula Weight 324.0998
Chemical Name or Material 6-Hydroxy Warfarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.